missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RII/RIII (CD32/CD16) Antibody (2.4G2) - Azide and BSA Free, Novus Biologicals™

Código de producto. 18667087 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.2 mg
Unit Size:
0.20 miligramo
25 microgramos
Product Code. Cantidad unitSize
18667087 0.025 mg 25 microgramos
18660787 0.2 mg 0.20 miligramo
2 options
This item is not returnable. View return policy

Product Code. 18667087

Brand: Novus Biologicals NBP3089840.025mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rat Monoclonal Antibody

Fc gamma RII/RIII (CD32/CD16) Monoclonal antibody specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Fc gamma RII/RIII (CD32/CD16)
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Neutralization
Clasificación Monoclonal
Clon 2.4G2
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS
Alias de gen CD16;CD16A;CD16-II;FCG3;FCGR3;FCGR3A;FCGRIII;FcGRIIIA;FCR-10;FCRIII;FCRIIIA;IGFR3;IMD20;Low affinity immunoglobulin gamma Fc region receptor III-A
Especie del huésped Rat
Inmunógeno Mouse BALB/c Macrophage J774 Cell Line.
Método de purificación Protein A purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Immunology
Primario o secundario Primary
ID de gen (Entrez) 2214
Especies diana Mouse
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.