Description
FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, RNA Inhibition, Knockout Validated, Knockdown Validated.
Specifications
Specifications
| Dilución | Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated |
| Alias de gen | DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 |
| Símbolos de los genes | FANCD2 |
| Especies diana | Human, Mouse, Canine, Kangaroo |
| Contenido y almacenamiento | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Isotype | IgG |
For Research Use Only