Learn More
FANCD2 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18785473 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
Tamaño de la unidad:
0.05 ml
25 microlitros
18785473 0.025 mL 25 microlitros
18787173 - 0.05 ml
2 options
Código de producto. 18785473
Marca: Novus Biologicals NB100182SS
Rabbit Polyclonal Antibody has been used in 178 publications
FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, RNA Inhibition, Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno FANCD2
Aplicaciones Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Formulación PBS with 0.05% Sodium Azide
N.º de referencia del gen Q9BXW9
Alias de gen DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Símbolos de los genes FANCD2
Especie del huésped Rabbit
Inmunógeno This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Peso molecular del antígeno 164.1 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primario o secundario Primary
ID de gen (Entrez) 2177
Especificidad de la prueba This antibody is specific for FANCD2.
Especies diana Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish
Contenido y almacenamiento Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only