missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
FAM156A Polyclonal antibody specifically detects FAM156A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | FAM156A |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulación | PBS (pH 7.2), 40% Glycerol |
| N.º de referencia del gen | Q8NDB6 |
| Alias de gen | family with sequence similarity 156, member A, TMEM29 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against Recombinant Protein corresponding to amino acids: MDPLQKRNPASPSKSSPMTAAETSQEGPAPSQPSYSEQPMMGLSNLSPGPGPSQAVPLPEGLLRQRYREEKTLEE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?