missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
FAM111B Polyclonal specifically detects FAM111B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
| Antígeno | FAM111B |
| Aplicaciones | Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | DyLight 594 |
| Dilución | Immunocytochemistry/Immunofluorescence |
| Formulación | 50mM Sodium Borate |
| Alias de gen | family with sequence similarity 111, member B |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with E. coli-derived Human FAM111B fragment. |
| Método de purificación | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?