Learn More
FADS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14118-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.Especificaciones
| FADS | |
| Polyclonal | |
| Unconjugated | |
| FLAD1 | |
| FAD pyrophosphorylase, FAD synthase, FAD1, FADS, FLAD1, FMN adenylyltransferase, PP591, RP11 307C12.7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319945, 751787, 80308 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8NFF5, Q8R123 | |
| Flad1 | |
| FADS Fusion Protein Ag5267 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |