missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII Antibody (RFF-VIII:c/8), Alexa Fluor™ 488, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Factor VIII Monoclonal specifically detects Factor VIII in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
| Antígeno | Factor VIII |
| Aplicaciones | Western Blot, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | RFF-VIII:c/8 |
| Conjugado | Alexa Fluor 488 |
| Dilución | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component |
| Símbolos de los genes | F8 |
| Especie del huésped | Mouse |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?