missing translation for 'onlineSavingsMsg'
Learn More

F-Spondin/SPON1 Antibody, Novus Biologicals™

Product Code. 18515714 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.02 mg
0.025 mg
Unit Size:
20 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18515714 0.02 mg 20 microgramos
18513123 0.025 mg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18515714 Supplier Novus Biologicals Supplier No. NB3003390.02mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chicken Polyclonal Antibody

F-Spondin/SPON1 Polyclonal specifically detects F-Spondin/SPON1 in Human, Mouse, Rat samples. It is validated for Western Blot, ICC/IF (Negative).
TRUSTED_SUSTAINABILITY

Specifications

Antígeno F-Spondin/SPON1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.91 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:200 - 1:500, ICC/IF (Negative)
Formulación PBS with 0.02% Sodium Azide
Alias de gen f-spondin, KIAA0762VSGP/F-spondin, MGC10724, spondin 1, (f-spondin) extracellular matrix protein, spondin 1, extracellular matrix protein, spondin-1, Vascular smooth muscle cell growth-promoting factor, VSGP
Símbolos de los genes SPON1
Especie del huésped Chicken
Inmunógeno Synthetic peptide, corresponding to amino acids 549-807 of SPON1.
Método de purificación Affinity Purified
Cantidad 0.02 mg
Estado normativo RUO
Disciplina de investigación Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 10418
Especificidad de la prueba SPON1
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.