missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etil 3-(trifluorometil)benzoato, 98 %, Thermo Scientific™
65.60€ - 292.00€
Identificadores de productos químicos
CAS | 76783-59-0 |
---|---|
Fórmula molecular | C10H9F3O2 |
Peso molecular (g/mol) | 218.175 |
Número MDL | MFCD00013560 |
Clave InChI | MHNBTKIAHHECCQ-UHFFFAOYSA-N |
Sinónimo | ethyl 3-trifluoromethyl benzoate, 3-trifluoromethyl benzoic acid ethyl ester, ethyl m-trifluoromethylbenzoate, ethyl-3-trifluoromethyl benzoate, ethyl 3-trifluoromethyl-benzoate, benzoic acid, 3-trifluoromethyl-, ethyl ester, ksc496o4j, ethyl a,a,a-trifluoro-m-toluate, 3-trifluoromethylbenzoic acid, ethyl ester |
PubChem CID | 144790 |
Nombre IUPAC | Etil 3-(trifluorometil)benzoato |
SMILES | CCOC(=O)C1=CC(=CC=C1)C(F)(F)F |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
76783-59-0 | |
218.175 | |
MHNBTKIAHHECCQ-UHFFFAOYSA-N | |
144790 | |
CCOC(=O)C1=CC(=CC=C1)C(F)(F)F |
C10H9F3O2 | |
MFCD00013560 | |
ethyl 3-trifluoromethyl benzoate, 3-trifluoromethyl benzoic acid ethyl ester, ethyl m-trifluoromethylbenzoate, ethyl-3-trifluoromethyl benzoate, ethyl 3-trifluoromethyl-benzoate, benzoic acid, 3-trifluoromethyl-, ethyl ester, ksc496o4j, ethyl a,a,a-trifluoro-m-toluate, 3-trifluoromethylbenzoic acid, ethyl ester | |
Etil 3-(trifluorometil)benzoato |
Especificaciones
Ethyl 3-(trifluoromethyl)benzoate | |
100°C to 101°C (10 mmHg) | |
1.446 | |
1968226 | |
MHNBTKIAHHECCQ-UHFFFAOYSA-N | |
Etil 3-(trifluorometil)benzoato | |
144790 | |
98% |
76783-59-0 | |
C10H9F3O2 | |
MFCD00013560 | |
ethyl 3-trifluoromethyl benzoate, 3-trifluoromethyl benzoic acid ethyl ester, ethyl m-trifluoromethylbenzoate, ethyl-3-trifluoromethyl benzoate, ethyl 3-trifluoromethyl-benzoate, benzoic acid, 3-trifluoromethyl-, ethyl ester, ksc496o4j, ethyl a,a,a-trifluoro-m-toluate, 3-trifluoromethylbenzoic acid, ethyl ester | |
CCOC(=O)C1=CC(=CC=C1)C(F)(F)F | |
218.175 | |
218.18 |
Seguridad y manipulación
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : Yes