Learn More
2-Oxobutirato de etilo, 90 %, Thermo Scientific Chemicals
16.60€ - 54.80€
Identificadores de productos químicos
CAS | 15933-07-0 |
---|---|
Fórmula molecular | C6H10O3 |
Peso molecular (g/mol) | 130.14 |
Número MDL | MFCD03265336 |
Clave InChI | FJAKCEHATXBFJT-UHFFFAOYSA-N |
Sinónimo | butanoic acid, 2-oxo-, ethyl ester, ethyl 2-ketobutyrate, 2-oxo-butyric acid ethyl ester, ethyl 3-methylpyruvate, ethyl a-oxobutyrate, ethyl2-oxobutanoate, ethyl 2-oxobutyrate, ethyl a-ketobutyrate, pubchem24313, ethyl-.alpha.-ketobutyrate |
PubChem CID | 85195 |
Nombre IUPAC | 2-oxobutanoato de etilo |
SMILES | CCOC(=O)C(=O)CC |
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
15933-07-0 | |
130.14 | |
FJAKCEHATXBFJT-UHFFFAOYSA-N | |
85195 | |
CCOC(=O)C(=O)CC |
C6H10O3 | |
MFCD03265336 | |
butanoic acid, 2-oxo-, ethyl ester, ethyl 2-ketobutyrate, 2-oxo-butyric acid ethyl ester, ethyl 3-methylpyruvate, ethyl a-oxobutyrate, ethyl2-oxobutanoate, ethyl 2-oxobutyrate, ethyl a-ketobutyrate, pubchem24313, ethyl-.alpha.-ketobutyrate | |
2-oxobutanoato de etilo |
Especificaciones
Ethyl 2-oxobutyrate | |
Amarillo | |
MFCD03265336 | |
Light sensitive | |
FJAKCEHATXBFJT-UHFFFAOYSA-N | |
2-oxobutanoato de etilo | |
85195 | |
90% |
15933-07-0 | |
C6H10O3 | |
1753936 | |
butanoic acid, 2-oxo-, ethyl ester, ethyl 2-ketobutyrate, 2-oxo-butyric acid ethyl ester, ethyl 3-methylpyruvate, ethyl a-oxobutyrate, ethyl2-oxobutanoate, ethyl 2-oxobutyrate, ethyl a-ketobutyrate, pubchem24313, ethyl-.alpha.-ketobutyrate | |
CCOC(=O)C(=O)CC | |
130.14 | |
130.14 | |
Líquido |
Seguridad y manipulación
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 240-071-7
missing translation for 'tsca' : No
missing translation for 'storageNote' : De - 30 °C a - 10 °C
Recommended Storage : Almacenar a - 20 °C
RUO – Research Use Only