Learn More
Invitrogen™ Human ERR Beta, Ligand Binding Domain, GST Tag Recombinant Protein
Código de producto. 10309164
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
Código de producto. 10309164
Marca: Invitrogen™ PV4802
Recombinant Protein
Estrogen Related Receptor beta, Ligand Binding Domain (ERR beta-LBD) is a 54.29 kDa recombinant human protein (amino acids 204-433) expressed as a GST fusion protein in baculovirus-infected insect cells. ERR beta was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies. Binding Domain, GST-Tagged, Recombinant Human Protein Form: Ligand Binding Domain
ESRRB (steroid hormone receptor ERR2) plays a role in maintenance of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survival of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso O95718
Concentración See Label
Para utilizar con (aplicación) Ligand Binding Assay
Formulación proprietary buffer with no preservative
ID de gen (Entrez) 2103
Nombre Human ERR Beta, Ligand Binding Domain, GST Tag
Cantidad 50 μg
Estado normativo RUO - research use only
Alias de gen DFNB35; ERR beta-2; ERR2; Err-2; ERRb; ERRB2; ERRbeta; ERR-beta; ERRbeta-2; ESRL2; ESRRB; estrogen receptor related 2; estrogen receptor-like 2; estrogen related receptor beta; estrogen related receptor, beta; estrogen-related nuclear receptor beta; estrogen-related receptor beta; estrogen-related receptor, beta; Estrrb; NR3B2; nuclear receptor ERRB2; nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; steroid hormone receptor ERR2
Nombre común ESRRB
Longitud de la proteína 204-433
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Sistema de expresión Baculovirus
Formulario Liquid
Subtipo de proteína Hormone Receptors