missing translation for 'onlineSavingsMsg'
Learn More

ERK2 Antibody, Novus Biologicals™

Product Code. 18209150 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
Product Code. Cantidad unitSize
18209150 100 μL 100 microlitros
1 options
This item is not returnable. View return policy

Product Code. 18209150

Brand: Novus Biologicals NBP119102

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ERK2 Polyclonal specifically detects ERK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ERK2
Aplicaciones Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500
Formulación Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
N.º de referencia del gen NP_002736.3
Alias de gen EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Símbolos de los genes MAPK1
Especie del huésped Rabbit
Inmunógeno The immunogen recognized by this antibody maps to a region between residue 310 and 360 of human extracellular signal-regulated kinase-2 (mitogen-activated protein kinase 1) using the numbering given in entry NP_002736.3 (GeneID 5594).
Método de purificación Affinity Purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Signal Transduction, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 5594
Especificidad de la prueba Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee,Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.