missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The ERG28 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
| Línea de células | The lysate was created in HEK293T cells, using plasmid ID RC2238 and based on accession number NM_7176. The protein contains a C-terminal DDK tag. |
| Para utilizar con (aplicación) | Western Blot |
| Cantidad | 0,1 mg |
| Tampón de almacenamiento | RIPA buffer |
| Pureza | Protein |
| Especificidad de la prueba | Homo sapiens chromosome 14 open reading frame 1 (C14orf1), mRNA. |
| Contenido y almacenamiento | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Para uso exclusivo en investigación
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?