Learn More
Novus Biologicals™ ErbB3/Her3 Antibody (patritumab), Novus Biologicals™
Código de producto. 30048141 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30048141 1 mg 1 miligramo
30045949 50 μg 50 microgramos
30047947 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30048141 Proveedor Novus Biologicals™ N.º de proveedor NBP3281841mg
Human Monoclonal Antibody
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno ErbB3/Her3
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon patritumab
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen P21860
Alias de gen c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Especie del huésped Human
Inmunógeno ERBB3/ HER3
Método de purificación Protein A purified
Cantidad 1 mg
Estado normativo RUO
Disciplina de investigación Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 2065
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1