Learn More
Ephrin-A1 Antibody (CL6501) [PE], Novus Biologicals™
Código de producto. 30297653 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30297653 0.1 mL 0.10 ml
1 options
Código de producto. 30297653 Proveedor Novus Biologicals N.º de proveedor NBP276497PE
Mouse Monoclonal Antibody
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Ephrin-A1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon CL6501
Conjugado PE
Formulación PBS
Alias de gen ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P20827, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer
Primario o secundario Primary
ID de gen (Entrez) 1942
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1