Learn More
Novus Biologicals™ EphA2 Antibody (Sanofi Aventis patent anti-EphA2) - Humanized, Novus Biologicals™
Código de producto. 30044602 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30044602 50 μg 50 microgramos
30045684 100 μg 100 microgramos
30048236 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30044602 Proveedor Novus Biologicals™ N.º de proveedor NBP32835450ug
Human Monoclonal Antibody
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno EphA2
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon Sanofi Aventis patent anti-EphA2
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen P29317
Alias de gen ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Especie del huésped Human
Inmunógeno EphA2
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cancer, Protein Kinase
Primario o secundario Primary
ID de gen (Entrez) 1969
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1