Learn More
Epcam / CD326 Rabbit, Clone: EGP40/1556R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 4072-RBM16-P1
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| Epcam / CD326 | |
| Cell surface | |
| EGP40/1556R | |
| Unconjugated | |
| NP_002345 | |
| Rabbit | |
| 40-43kDa | |
| Protein A/G Purified | |
| RUO | |
| 4072 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| EPCAM | |
| Recombinant human EPCAM protein | |
| HT29 cells or breast tumor | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |