Learn More
EPAC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12572-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The EPAC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPAC1. This antibody recognizes human antigen. The EPAC1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
| EPAC1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 10411 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 540 μg/mL | |
| Rabbit | |
| Primary | |
| Human |