missing translation for 'onlineSavingsMsg'
Learn More

Endothelial Lipase Antibody, FITC, Novus Biologicals™

Código de producto. 18230080 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18230080 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18230080

Brand: Novus Biologicals NB400111F

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Endothelial Lipase
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Clasificación Polyclonal
Conjugado FITC
Dilución Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulación PBS with 0.05% Sodium Azide
N.º de referencia del gen Q9Y5X9
Alias de gen EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Símbolos de los genes LIPG
Especie del huésped Rabbit
Inmunógeno A C-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Unfolded Protein Response
Primario o secundario Primary
ID de gen (Entrez) 9388
Especificidad de la prueba Human. Predicted to react with monkey based on 100% sequence homology. Does not appear to recognize mouse.
Especies diana Human, Mouse (Negative)
Contenido y almacenamiento Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.