missing translation for 'onlineSavingsMsg'
Learn More

Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Antibody (A7L6), Novus Biologicals™

Código de producto. p-5541196 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mg
0.1 mg
Tamaño de la unidad:
0.02 miligramo
0.10 miligramo
Código de producto. Cantidad unitSize
18160779 0.02 mg 0.02 miligramo
18180789 0.1 mg 0.10 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18160779

Marca: Novus Biologicals NBP2444480.02MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Monoclonal specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human, Mouse, Porcine, Bovine, Fish, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Endorepellin/Perlecan/Heparan Sulfate Proteoglycan
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Clasificación Monoclonal
Clon A7L6
Concentración 0.2mg/mL
Conjugado Unconjugated
Dilución Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Formulación 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
N.º de referencia del gen P98160
Alias de gen basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1
Símbolos de los genes HSPG2
Especie del huésped Rat
Inmunógeno Murine EHS laminin preparation
Método de purificación Protein A or G purified
Cantidad 0.02 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3339
Especificidad de la prueba This MAb specifically precipitates heterogeneous material of high MW, identified as perlecan, a major heparan-sulfate proteoglycan (HSPG) within all basement membranes and cell surfaces. It does not cross-react with laminin, fibronectin, or dermatran sulfate proteoglycan. Because of perlecan s strategic location and ability to store and protect growth factors, it has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a co-receptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.
Especies diana Human, Mouse, Pig, Fish, Monkey
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG2a κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.