missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endocan Monoclonal Antibody (C7), Invitrogen™

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA543841
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Endothelial cell-specific molecule 1 is a protein that in humans is encoded by the ESM1 gene. This gene encodes a secreted protein which is mainly expressed in the endothelial cells in human lung and kidney tissues. The expression of this gene is regulated by cytokines, suggesting that it may play a role in endothelium-dependent pathological disorders. The transcript contains multiple polyadenylation and mRNA instability signals. Two transcript variants encoding different isoforms have been found for this gene.
Especificaciones
| Endocan | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9NQ30 | |
| ESM1 | |
| Trp20-Arg184 (Accession Q9NQ30), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C7 | |
| Unconjugated | |
| ESM1 | |
| 0610042H23Rik; AV004503; endocan; endothelial cell specific molecule 1; endothelial cell-specific molecule 1; ESM1; ESM-1; ESM-1 secretory protein; IGFBP-rp6; PG25; pineal specific PG25 protein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido