Descripción
Elp4 Monoclonal antibody specifically detects Elp4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | Elp4 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | SR1794 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | C11orf19, chromosome 11 open reading frame 19, dJ68P15A.1, elongation protein 4 homolog (S. cerevisiae), elongator complex protein 4, hELP4, PAX6 neighbor gene protein, PAX6NEB, PAXNEBFLJ20498 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human Elp4 (Uniprot #: Q96EB1) |
| Mostrar más |