missing translation for 'onlineSavingsMsg'
Learn More

Elastin Antibody, Novus Biologicals™

Código de producto. 30230307 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Unit Size:
100 microlitros
20 microlitros
Product Code. Cantidad unitSize
30230307 20 μL 20 microlitros
30232137 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 30230307

Brand: Novus Biologicals NBP33801020ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Elastin Polyclonal antibody specifically detects Elastin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Elastin
Aplicaciones ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulación PBS (pH 7.3), 50% glycerol
Alias de gen elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 27-200 of human Elastin (NP_001265868.1).,, Sequence:, GGVPGAIPGGVPGGVFYPGAGLGALGGGALGPGGKPLKPVPGGLAGAGLGAGLGAFPAVTFPGALVPGGVADAAAAYKAAKAGAGLGGVPGVGGLGVSAGAVVPQPGAGVKPGKVPGVGLPGVYPGGVLPGARFPGVGVLPGVPTGAGVKPKAPGVGGAFAGIPGVGPFGGPQP
Método de purificación Affinity purified
Cantidad 20 μL
Estado normativo RUO
Disciplina de investigación Extracellular Matrix
Primario o secundario Primary
ID de gen (Entrez) 2006
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.