Learn More
Elafin/Skalp Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15963-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Neutrophil and pancreatic elastase-specific inhibitor of skin. It may prevent elastase-mediated tissue proteolysis.Especificaciones
| Elafin/Skalp | |
| Polyclonal | |
| Unconjugated | |
| PI3 | |
| cementoin, Elafin, Elastase specific inhibitor, ESI, Peptidase inhibitor 3, PI 3, PI3, Protease inhibitor WAP3, SKALP, WAP3, WFDC14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5266 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P19957 | |
| PI3 | |
| Elafin/Skalp Fusion Protein Ag8735 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |