missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
eIF4A2 Monoclonal antibody specifically detects eIF4A2 in Human samples. It is validated for ELISA, Protein Array
Specifications
Specifications
| Antígeno | eIF4A2 |
| Aplicaciones | ELISA, Protein Array |
| Clasificación | Monoclonal |
| Clon | PCRP-EIF4A2-2B5 |
| Conjugado | Alexa Fluor 350 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant full-length human eIF4A2 protein (Uniprot: Q14240) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?