missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
eIF3K Monoclonal antibody specifically detects eIF3K in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antígeno | eIF3K |
| Aplicaciones | Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 01 |
| Conjugado | Unconjugated |
| Dilución | Immunocytochemistry/ Immunofluorescence 1:20-1:100 |
| Formulación | 0.2 um filtered solution in PBS |
| Alias de gen | ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human eIF3K (Uniprot#: Q9UBQ5; Ala2-Gln218). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?