missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EGFR (ErbB1) [T790M, L858R], GST Tag Recombinant Protein

Código de producto. 10479793
Change view
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10479793 10 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10479793 Proveedor Invitrogen™ N.º de proveedor PV4879

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic.

EGFR, epidermal growth factor receptor, is a receptor tyrosine kinases that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. EGFR is encoded by the EGFR gene located on chromosome 7 in humans. EGFR belongs to the HER/ERbB family of proteins that includes three other receptor tyrosine kinases, ERbB2, ERbB3, ERbB4. EGFR is a transmembrane receptor and binding of its cognate ligands such as EGF (Epidermal Growth Factor) and TGF alpha (Transforming Growth Factor alpha) to the extracellular domain leads to EGFR dimerization followed by autophosphorylation of the tyrosine residues in the cytoplasmic domain. Overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.
TRUSTED_SUSTAINABILITY

Especificaciones

Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris with 0.02% polysorbate 20, 50% glycerol, 2mM DTT, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Peso molecular 90.6 kDa
Nombre Human EGFR (ErbB1) [T790M, L858R], GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Nombre común EGFR (T790M, L858R Mutant)
Familia Quinasa EGF Receptor (ErbB) Kinase Family
Longitud de la proteína 668-1210; T790M, L858R
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia 668-1210; T790M, L858R
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical binding/kinetic
Grupo de cinasas Tyrosine Kinases
Condiciones de envío Dry Ice
Compatibilidad de sistemas Z'-LYTE™
Entrada diana EGFR T790M L858R
Gen diana Epidermal growth factor receptor
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Catalytic Domain, Cytoplasmic, Mutant
Formulario Liquid
Subtipo de proteína Receptor Tyrosine Kinases
Categoría de investigación Oncology, Signal Transduction
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.