Learn More
Novus Biologicals™ EGFR Antibody (Duke D2C7), Novus Biologicals™
Código de producto. 30047697 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30047697 1 mg 1 miligramo
30048204 50 μg 50 microgramos
30048414 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30047697 Proveedor Novus Biologicals™ N.º de proveedor NBP3284641mg
Human Monoclonal Antibody
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno EGFR
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon Duke D2C7
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen P00533
Alias de gen avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Especie del huésped Human
Inmunógeno ERBB1/ EGFR/ HER1
Método de purificación Protein A purified
Cantidad 1 mg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
Primario o secundario Primary
ID de gen (Entrez) 1956
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1