missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
ECT2 Polyclonal antibody specifically detects ECT2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | ECT2 |
| Aplicaciones | Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Formulación | PBS (pH 7.2) and 40% Glycerol |
| Alias de gen | ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to amino acids: SLKEVMTHINEDKRKTEAQKQIFDVVYEVDGCPANLLSSHRSLVQRVETISLGEHPCDRGEQVTLFLFNDCLEIARKRHKVIGTFRSPH |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?