Learn More
ECSIT Antibody, Novus Biologicals™
Código de producto. p-200066642 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
Tamaño de la unidad:
25 microgramos
18561924 0.025 mg 25 microgramos
1 options
Código de producto. 18561924
Marca: Novus Biologicals NBP1765940.025mg
Rabbit Polyclonal Antibody
ECSIT Polyclonal specifically detects ECSIT in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno ECSIT
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen NP_057665
Alias de gen ECSIT homolog (Drosophila), evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial, likely ortholog of mouse signaling intermediate in Toll pathway evolutionarilyconserved, Protein SITPEC, signaling intermediate in Toll pathway evolutionarily conserved ortholog, SITPECsignaling intermediate in Toll pathway, evolutionarily conserved
Símbolos de los genes ECSIT
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human ECSIT. The immunogen is located within amino acids 290 - 340 of ECSIT. Amino Acid Squence: PPEEREVEETPEEW
Método de purificación Ion exchange chromatography
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 51295
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only