missing translation for 'onlineSavingsMsg'
Learn More

ECSIT Antibody, Novus Biologicals™

Product Code. 18561924 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.025 mg
Unit Size:
25 microgramos
Product Code. Cantidad unitSize
18561924 0.025 mg 25 microgramos
1 options
This item is not returnable. View return policy

Product Code. 18561924

Brand: Novus Biologicals NBP1765940.025mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ECSIT Polyclonal specifically detects ECSIT in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ECSIT
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen NP_057665
Alias de gen ECSIT homolog (Drosophila), evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial, likely ortholog of mouse signaling intermediate in Toll pathway evolutionarilyconserved, Protein SITPEC, signaling intermediate in Toll pathway evolutionarily conserved ortholog, SITPECsignaling intermediate in Toll pathway, evolutionarily conserved
Símbolos de los genes ECSIT
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human ECSIT. The immunogen is located within amino acids 290 - 340 of ECSIT. Amino Acid Squence: PPEEREVEETPEEW
Método de purificación Ion exchange chromatography
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 51295
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.