missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human samples. It is validated for Western Blot, ELISA
Especificaciones
Especificaciones
| Antígeno | EAAT2/GLT1 |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Monoclonal |
| Clon | 4G8 |
| Conjugado | Unconjugated |
| Dilución | Western Blot, ELISA |
| Formulación | In 1x PBS, pH 7.4 |
| N.º de referencia del gen | NP_004162 |
| Alias de gen | GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 |
| Especie del huésped | Mouse |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?