Learn More
E6AP/UBE3A Antibody, mFluor Violet 610 SE, Novus Biologicals™
Código de producto. 18588191 Tienda Bio Techne Productos
Código de producto. 18588191
Marca: Novus Biologicals NBP299342MFV610
Rabbit Polyclonal Antibody
E6AP/UBE3A Polyclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)