missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Doublecortin Recombinant Rabbit Monoclonal Antibody (BLR168J)

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA544521
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.
Especificaciones
| Doublecortin | |
| Recombinant Monoclonal | |
| 50 μg/mL | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
| O43602 | |
| DCX | |
| Peptide representing a region between residue 200 and 250 of Human Doublecortin. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| BLR168J | |
| Unconjugated | |
| DCX | |
| DBCN; Dbct; DC; Dcn; DCX; doublecortex; doublecortin; doublin; lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS | |
| Rabbit | |
| purified | |
| RUO | |
| 1641 | |
| 4°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido