Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 488 (Pre-adsorbed), Novus Biologicals™
Código de producto. 18775923 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
0.10 miligramo
18775923 0.1 mg 0.10 miligramo
1 options
Código de producto. 18775923
Marca: Novus Biologicals NBP172966
Donkey Polyclonal Antibody
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IgG (H+L)
Aplicaciones Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ELISA
Clasificación Polyclonal
Concentración LYOPH
Conjugado DyLight 488
Dilución Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Dot Blot, Fluorophore-linked immunosorbent assay 1:20000
Especie del huésped Donkey
Inmunógeno Rabbit IgG whole molecule
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Immunology
Primario o secundario Secondary
Especificidad de la prueba This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat and Sheep Serum Proteins. This antibody will react with heavy chains of rabbit IgG and with light chains of most rabbit immunoglobulins.
Reconstitución Reconstitute with 100 ul deionized water (or equivalent).
Especies diana Rabbit
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only