Learn More
DNM2 Mouse, anti-Human, CoraLite 555, Clone: 1B8H3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL555-68209-20UL
Descripción
The DNM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DNM2. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The DNM2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™555.
Especificaciones
| DNM2 | |
| 1B8H3 | |
| CoraLite 555 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| CMTDI1, CMTDIB, DI CMTB, DNM2, DYN2, dynamin 2, DYNII | |
| 20 μL | |
| 1785 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.