Learn More
DNM2 Mouse, anti-Human, CoraLite 555, Clone: 1B8H3, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL555-68209-20UL
 Ver más versiones de este producto
Código de producto. 18005395
  • 140.00€ / 20 microlitros
Descripción
The DNM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DNM2. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The DNM2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™555.
Especificaciones
Especificaciones
DNM2
1B8H3
CoraLite 555
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
CMTDI1, CMTDIB, DI CMTB, DNM2, DYN2, dynamin 2, DYNII
20 μL
1785
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.