missing translation for 'onlineSavingsMsg'
Learn More

DNAL1 Antibody (29E4), DyLight 350, Novus Biologicals™

Código de producto. 18157468 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18157468 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18157468 Proveedor Novus Biologicals N.º de proveedor NBP242657UV

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNAL1 Monoclonal specifically detects DNAL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno DNAL1
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 290000
Conjugado DyLight 350
Dilución Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen C14orf168MGC12435,1700010H15RiK, chromosome 14 open reading frame 168, dynein light chain 1, axonemal, dynein, axonemal, light chain 1
Símbolos de los genes DNAL1
Especie del huésped Mouse
Inmunógeno Recombinant human DNAL1 (1-190aa) purified from E. coli
Método de purificación Protein A purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 83544
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.