missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21112-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.Especificaciones
| DKK1 | |
| Polyclonal | |
| Unconjugated | |
| DKK1 | |
| Dickkopf 1, Dickkopf related protein 1, DKK 1, DKK1, hDkk 1, SK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13380, 22943, 293897 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54908, O94907 | |
| Dkk1 | |
| DKK1 Fusion Protein Ag15110 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido