Learn More
DIS3L2 Mouse anti-Human, Unconjugated, Clone: 2C9F2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67623-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The DIS3L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DIS3L2. This antibody recognizes human antigen. The DIS3L2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Especificaciones
| DIS3L2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Flow Cytometry, ELISA | |
| 2C9F2 | |
| Unconjugated | |
| 150 μL | |
| 129563 | |
| IgG1 |