Learn More
Dihydrofolate Reductase/DHFR Antibody (OTI6G7), Allophycocyanin, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | Dihydrofolate Reductase/DHFR |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | OTI6G7 |
| Conjugado | APC |
| Dilución | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Formulación | PBS |
| Alias de gen | DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 |
| Especie del huésped | Mouse |
| Inmunógeno | Full-length protein expressed in 293T cell transfected with human DHFR expression vector |
| Mostrar más |