missing translation for 'onlineSavingsMsg'
Learn More

DIAPH1 Antibody, HRP, Novus Biologicals™

Código de producto. 18590631 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18590631 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18590631

Marca: Novus Biologicals NB600231H

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIAPH1 Polyclonal antibody specifically detects DIAPH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno DIAPH1
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado HRP
Dilución Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulación PBS
Alias de gen DIA1, diaphanous homolog 1 (Drosophila), hDIA1, homolog) 1, LFHL1FLJ25265
Especie del huésped Rabbit
Inmunógeno The immunogen recognized by this antibody maps to a region between residues 1200 and the C-terminus (residue 1248) of human Diaphanous Homolog 1 using the numbering given in entry NP_005210.1 (GeneID 1729).
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 1729
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.