Learn More
DHRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15393-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.Especificaciones
| DHRS3 | |
| Polyclonal | |
| Unconjugated | |
| DHRS3 | |
| DD83.1, DHRS3, RDH17, retSDR1, Rsdr1, SDR1, SDR16C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20148, 313689, 9249 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75911, O88876 | |
| Dhrs3 | |
| DHRS3 Fusion Protein Ag7589 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |