missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Destrin Monoclonal antibody specifically detects Destrin in Canine,Human,Monkey,Mouse,Primate samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | Destrin |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | OTI2F7 |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | ACTDPdestrin, Actin-depolymerizing factor, ADFbA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), bA462D18.2, destrin (actin depolymerizing factor), DSN |
| Especie del huésped | Mouse |
| Inmunógeno | Full length human recombinant protein of human DSTN (NP_006861) produced in HEK293T cell. |
| Método de purificación | Immunogen affinity purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?