missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Desmocollin-1 Polyclonal antibody specifically detects Desmocollin-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | Desmocollin-1 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulación | PBS (pH 7.2) and 40% Glycerol |
| Alias de gen | cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, desmosomal glycoprotein 2/3, DG2/DG3 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to amino acids: RQVILQVGVINEAQFSKAASSQTPTMCTTTVTVKIIDSDEGPECHPPVKVIQSQDGFPAGQELLGYKALDPEISSGEGLRYQKLGDEDNWFEINQHTG |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?