missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dectin-1/CLEC7A Antibody (Baylor patent anti-Dectin-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28458-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Dectin-1/CLEC7A Monoclonal antibody specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
| Dectin-1/CLEC7A | |
| Monoclonal | |
| Unconjugated | |
| Q9BXN2 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| Baylor patent anti-Dectin-1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| CLEC7A | |
| 1 mg | |
| Signal Transduction | |
| 64581 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido