missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Decorin Neo Polyclonal specifically detects Decorin Neo in Human, Mouse, Bovine samples. It is validated for Western Blot.
Specifications
Specifications
| Antígeno | Decorin Neo |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 2 μg/mL |
| Formulación | PBS containing 1 mg/ml BSA with 0.05% Sodium Azide |
| Alias de gen | Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B |
| Símbolos de los genes | DCN |
| Especie del huésped | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?