missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
44531 Polyclonal specifically detects 44531 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing.
Especificaciones
Especificaciones
| Antígeno | DEC1 |
| Aplicaciones | Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:5000-1:15000, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 29715265), Flow Cytometry 2-5 ug/ml, ELISA reported in scientific literature (PMID 31061528), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 31061528) |
| Formulación | PBS with 0.02% Sodium Azide |
| N.º de referencia del gen | O14503 |
| Alias de gen | basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14 |
| Símbolos de los genes | BHLHE40 |
| Mostrar más |
For Research Use Only
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?