missing translation for 'onlineSavingsMsg'
Learn More

DEC-205/CD205 Antibody (019), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18549815 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μg
Unit Size:
100 microlitros
Product Code. Cantidad unitSize
18549815 100 μg 100 microlitros
1 options
This item is not returnable. View return policy

Product Code. 18549815

Brand: Novus Biologicals NBP312780AF405

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DEC-205/CD205 Monoclonal antibody specifically detects DEC-205/CD205 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno DEC-205/CD205
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 019
Conjugado Alexa Fluor 405
Formulación 50mM Sodium Borate
Alias de gen CD205;DCL-1 fusion;DEC-205;gp200;Ly-75;Lymphocyte antigen 75;membrane antigen MN;RP23-10L17.4
Especie del huésped Rabbit
Inmunógeno This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Mouse DEC-205/CD205
Método de purificación Protein A purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Cancer, Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primario o secundario Primary
Especies diana Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.