Descripción
DDX21 Polyclonal specifically detects DDX21 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).
Especificaciones
Especificaciones
| Clasificación | Polyclonal |
| Especie del huésped | Rabbit |
| Cantidad | 0.1 mL |
| Especies diana | Human, Mouse |
| Contenido y almacenamiento | Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| Isotype | IgG |
For Research Use Only