missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDB1 Mouse anti-Human, Mouse, Rat, Clone: 4F6C6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66010-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is specific to DDB1.
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.Especificaciones
| DDB1 | |
| Monoclonal | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16531, Q3U1J4, Q9ESW0 | |
| Ddb1 | |
| DDB1 Fusion Protein Ag17903 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 4F6C6 | |
| Unconjugated | |
| DDB1 | |
| DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 13194, 1642, 64470 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido